Learn More
Medchemexpress LLC HY-W050026 10mg Medchemexpress, Phenylacetylglutamine CAS:28047-15-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW05002610MG
Encompass_Preferred
Medchemexpress, HY-W050026 10mg Phenylacetylglutamine CAS:28047-15-6 Phenylacetylglutamine is a colonic microbial metabolite from amino acid fermentation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4662