Learn More
Selleck Chemical LLC Phenylacetylglutamine
SDP
Supplier:  Selleck Chemical LLC S607625MG
Encompass_Preferred
Phenylacetylglutamine is a product formed by the conjugation of phenylacetate and glutamine It is a normal constituent of human urine but other mammals such as the dog cat rat monkey sheep and horse do not excrete this compound
Catalog No. 50-000-04076