Learn More
Medchemexpress LLC Phenylacetylglycine | 500-98-1 | MFCD00021744 | 99.6% | 193.20 g·mol⁻1 | C10H11NO3 | 1 ML

Supplier: Medchemexpress LLC HYW01506110MM/1ML
Phenylacetylglycine is a gut microbial metabolite reported to activate β2-adrenergic receptor (β2AR) and to exhibit cardioprotective activity in ischemia/reperfusion models. Supplied as a ready-to-use solution for research, it is suitable for studies of endogenous metabolism, receptor signaling, and cardiac injury mechanisms.
- 10 mM solution in DMSO, 1 mL vial.
- High purity (≈99.6%) suitable for research use.
- Molecular formula C10H11NO3; molecular weight 193.20 g·mol⁻1.
- Active in β2-adrenergic receptor assays and metabolite studies.
- Convenient for in vitro experiments and signaling studies.
- Store at -20 °C according to supplier guidance.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.