Learn More
Medchemexpress LLC Phenylacetylglycine | 500-98-1 | MFCD00021744 | 99.6% | 193.20 g/mol | C10H11NO3 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW015061
Encompass_Preferred
Phenylacetylglycine is a gut microbial metabolite and analytical standard used in biochemical and pharmacology research. It has been reported to activate the β2-adrenergic receptor (β2AR) and to protect against cardiac injury caused by ischemia/reperfusion, making it useful for metabolic profiling, receptor pharmacology studies, and reference assays.
  • Used as an analytical standard for metabolic profiling and quantitation.
  • Reported activator of β2-adrenergic receptor, relevant in pharmacology studies.
  • Shown to protect against ischemia/reperfusion cardiac injury in studies.
  • Soluble in DMSO and compatible with common in vivo vehicles.
  • Stable as a powder under recommended frozen storage conditions.
Catalog No. 50-003-02360