Learn More
Medchemexpress LLC Phenylalanine, 4-chloro-, hydrochloride | 23633-07-0 | 98.6% | 236.10 | C9H11Cl2NO2 | 1g
SDP
Supplier:  Medchemexpress LLC HYW7728501G
Encompass_Preferred
Fenclonine hydrochloride is a selective and irreversible tryptophan hydroxylase inhibitor which is a rate-limiting enzyme in the biosynthesis of serotonin Fenclonine hydrochloride can be used in carcinoid syndrome research[1][2][3]
Catalog No. 50-002-68480