Learn More
Medchemexpress LLC Phenylalanine, 4-chloro-, methyl ester, hydrochloride | 14173-40-1 | MFCD00012530 | 99.9% | 250.12 | C10H13Cl2NO2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10145610MM 1ML
Encompass_Preferred
PCPA methyl ester hydrochloride (4-chloro-DL-phenylalanine methyl ester hydrochloride) is a reversible tryptophan hydroxylase inhibitor used to reduce serotonin (5-HT) synthesis in neuroscience research. It crosses the blood-brain barrier and is provided in solution and solid forms for in vitro and in vivo studies.
  • Reversible tryptophan hydroxylase inhibitor that reduces central 5-HT levels.
  • Crosses the blood-brain barrier, enabling central nervous system studies.
  • Available as a 10 mM solution in DMSO (1 mL) and as solid quantities for scaling experiments.
  • High purity (99.9%) suitable for research applications.
  • Molecular formula C10H13Cl2NO2; molecular weight 250.12.
Catalog No. 50-002-47099