Learn More
Medchemexpress LLC Phenylalanine, N-[(1,1-dimethylethoxy)carbonyl]-N,β,β-trimethyl- | 676487-35-7 | 99.1% | 307.39 | 1 ML

Supplier: Medchemexpress LLC HY4104510MM/1ML
(Rac)-Taltobulin intermediate-1 is an intermediate in the synthesis of Taltobulin. Taltobulin is a common toxin component in ADC preparation and a powerful tubulin inhibitor. It disrupts tubulin polymerization, induces mitotic arrest, and induces apoptosis.
- Intermediate in the synthesis of Taltobulin
- Common toxin component in ADC preparation
- Powerful tubulin inhibitor
- Disrupts tubulin polymerization
- Induces mitotic arrest
- Induces apoptosis
- Purity: 99.11%
- Appearance: Solid, white to off-white
- Solubility: DMSO : ≥ 100 mg/mL (325.32 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.