Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylarsine Oxide, Spectrum™ Chemical
SDP

$124.00 - $15510.00

Chemical Identifiers

CAS 637-03-6
Molecular Weight (g/mol) 168.03 g/mol
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
18-621-278
SDP
View Documents Promotion Details
Spectrum Chemical Manufacturing Corporation
P15801GM
1 g
Each for $124.00
Add to Cart
 
18-621-279
SDP
View Documents Promotion Details
Spectrum Chemical Manufacturing Corporation
P15805GM
5 g
Each for $327.00
Add to Cart
 
18-621-280
SDP
View Documents Promotion Details
Spectrum Chemical Manufacturing Corporation
P158025GM
25 g
Each for $1,325.00
Add to Cart
 
18-621-281
SDP
View Documents Promotion Details
Spectrum Chemical Manufacturing Corporation
P1580100GMBL
100 g
Each for $2,773.00
Add to Cart
 
18-621-282
SDP
View Documents Promotion Details
Spectrum Chemical Manufacturing Corporation
P15801KGBL
1 kg
Each for $15,510.00
Add to Cart
 
Description

Description

Specifications

Chemical Identifiers

637-03-6
168.03 g/mol

Specifications

637-03-6
168.03 g/mol
Amber Glass Bottle
Phenylarsine Oxide
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.