accessibility menu, dialog, popup

UserName

Viewing profile as user:
OAs

CAS RN 637-03-6

IUPAC Name: arsorosobenzene
Synonyms: oxophenylarsine
Molecular Weight (g/mol): 168.03
Molecular Formula: C6H5AsO
InChi Key: BQVCCPGCDUSGOE-UHFFFAOYSA-N
SMILES: O=[As]C1=CC=CC=C1
Quantity 
  • (10)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (4)
  • (7)
Molecular Weight (g/mol) 
  • (30)
  • (5)
Grade 
  • (9)
  • (4)
  • (13)
  • (1)
pH 
  • (2)
  • (9)
  • (3)
  • (1)
  • (4)
Physical Form 
  • (1)
  • (3)
  • (5)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (15)
  • (9)
  • (3)
  • (7)
  • (1)
  • (13)
  • (9)
  • (1)
  • (6)
  • (10)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (4)
  • (7)
  • (30)
  • (5)
  • (9)
  • (4)
  • (13)
  • (1)
  • (2)
  • (9)
  • (3)
  • (1)
  • (4)
  • (1)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)