Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenylbiguanide hydrochloride | 55-57-2 | MFCD00035040 | 99.8% | 213.67 g/mol | C8H12ClN5 | 10 G
SDP

Supplier:  Medchemexpress LLC HY101331A10G

Encompass_Preferred

Phenylbiguanide hydrochloride is the hydrochloride salt of phenylbiguanide, a selective 5-HT3 receptor agonist supplied as a high-purity research chemical for in vitro pharmacology and neuroscience studies. The material is characterized with standard identifiers, purity specification, and storage guidance for laboratory use.

  • Selective 5-HT3 receptor agonist with an EC50 of 3.0 ± 0.1 μM.
  • Chemical identifiers: CAS 55-57-2; formula C8H12ClN5; molecular weight 213.67 g/mol.
  • High purity solid (≈99.8%) suitable for analytical and pharmacological assays.
  • Available in multiple lab-scale pack sizes (10 G-100 G) for research workflows.
  • Storage: store at room temperature up to 3 years; in solvent, -80 °C (2 years) or -20 °C (1 year).
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-11259


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.