Learn More
Selleck Chemical LLC Phenylephrine HCl S2569-50mg
SDP
Supplier:  Selleck Chemical LLC S256950MG
Encompass_Preferred
Phenylephrine HCl is a selective 1-adrenergic receptor agonist used primarily as a decongestant
Catalog No. 50-311-1643