Learn More
Medchemexpress LLC Phenylephrine hydrochloride | 61-76-7 | 99.99% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0471500MG
Encompass_Preferred
Phenylephrine hydrochloride is a selective α1-adrenoceptor agonist, exhibiting pKis of 5.86, 4.87, and 4.70 for α1D, α1B, and α1A receptors respectively. It is intended for research use only.
  • Promotes cardiac fibroblast proliferation and activates CaN, evoking NFAT3 nuclear translocation.
  • Causes a rapid 12-fold activation of two p38-MAPK isoforms.
  • Increases heart contractility.
  • Activates SAPKs/JNKs in neonatal ventricular myocytes.
  • Increases alveolar fluid clearance in high tidal volume-ventilated rats and accelerates pulmonary edema absorption.
Catalog No. 50-004-07807