Learn More
Selleck Chemical LLC PhenylephrineHCl 10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S256910MM/1ML
Encompass_Preferred
Phenylephrine HCl (NCI-C55641) is a selective ?1-adrenergic receptor agonist, used primarily as a decongestant. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3088