Learn More
Medchemexpress LLC Phenylglyoxylic acid (Benzoylformic acid) | 611-73-4 | MFCD00002575 | 99.66% | 150.13 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01025510MM/1ML
Encompass_Preferred
Phenylglyoxylic acid, also known as Benzoylformic acid, is a metabolite of ethylbenzene and styrene (EB/S) and serves as a biomarker for exposure to these compounds in humans. It also functions as a drug intermediate in the synthesis of antineoplastic compounds. For research use only.
  • Biomarker for exposure to ethylbenzene and styrene.
  • Used in the synthesis of antineoplastic compounds.
  • Exhibits anti-inflammatory activity in animal models.
Catalog No. 50-004-51849