Learn More
Medchemexpress LLC Phenylmethan-d2-ol | 21175-64-4 | MFCD00084124 | 98.0% | 110.15 g/mol | C7H6D2O | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW450789S25MG
Encompass_Preferred
Phenylmethan-d2-ol is a deuterium-labeled analogue of benzyl alcohol (C7H6D2O) used as an isotope-labeled reference standard, tracer, or reagent in analytical and synthetic chemistry. It is suitable for mass spectrometry, mechanistic studies, and synthetic applications where a non-radioactive deuterium label is required. Supplied in milligram-scale quantities for laboratory use.
  • Deuterium-labeled benzyl alcohol (C7H6D2O) for isotope tracing.
  • High isotopic enrichment (~98 atom % D) suitable for mass spectrometry.
  • Ideal for mechanistic, kinetic, and tracer studies in synthesis and analysis.
  • Supplied in milligram-scale vials for small-scale laboratory experiments.
  • Non-radioactive stable label compatible with common analytical techniques.
Catalog No. 50-002-81067