Learn More
Medchemexpress LLC Phenylmethan-d2-ol | 21175-64-4 | MFCD00084124 | 99.0% | 110.15 | C7H6D2O | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW450789S50MG
Encompass_Preferred
Phenylmethan-d2-ol is the alpha,alpha-dideuterated form of benzyl alcohol used as a stable isotope reagent and internal standard in analytical and synthetic chemistry. It is supplied as a high-purity, isotopically enriched compound suitable for mass spectrometry, isotopic labeling, and mechanistic studies.
  • Deuterium label at alpha positions
  • High isotopic enrichment (>99.0%)
  • Molecular formula C7H6D2O, molecular weight 110.15
  • Small-quantity formats suitable for analytical workflows
  • Used as a mass spectrometry internal standard and isotopic tracer
  • Accompanied by product datasheet for purity and handling information
Catalog No. 50-002-81439