Learn More
Medchemexpress LLC Phenylmethyl 2-(acetylamino)-2-deoxy-α-D-glucopyranoside | 13343-62-9 | MFCD00070371 | ≥97.0% | 311.33 g/mol | C15H21NO6 | 1 G

Supplier: Medchemexpress LLC HYW0363881G
Phenylmethyl 2-(acetylamino)-2-deoxy-α-D-glucopyranoside is a benzyl glycoside reagent used in glycobiology and carbohydrate chemistry to study glycan structure, synthesis, enzymology, and protein-glycan recognition. It is supplied as a powder and intended for biochemical assay use with specified storage conditions to preserve stability.
- Used in glycobiology and carbohydrate chemistry assays.
- Powder form suitable for long-term storage.
- Purity ≥97.0% (NMR) ensures analytical quality.
- Molecular weight 311.33 g/mol and formula C15H21NO6 for reference.
- Storage: powder -20°C for 3 years or 4°C for 2 years; in solvent -80°C for 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.