Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenylmethyl 2-(acetylamino)-2-deoxy-α-D-glucopyranoside | 13343-62-9 | MFCD00070371 | 97.0% | 311.33 g·mol⁻¹ | C15H21NO6 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW0363885G

Encompass_Preferred

Phenylmethyl 2-(acetylamino)-2-deoxy-α-D-glucopyranoside is a benzyl-protected 2-acetamido-2-deoxy-α-D-glucopyranoside supplied for laboratory research in glycobiology and carbohydrate chemistry. The compound is provided as a powder and is characterized by CAS 13343-62-9, molecular formula C15H21NO6, and molecular weight 311.33 g·mol⁻¹. Recommended storage conditions and stability data are provided by the supplier.

  • Benzyl-protected 2-acetamido-2-deoxy-α-D-glucopyranoside for synthetic applications.
  • Supplied as a powder suitable for laboratory handling.
  • Molecular formula C15H21NO6 and molecular weight 311.33 g·mol⁻¹ for reference.
  • CAS registry number 13343-62-9 for unambiguous identification.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years).
  • In solvent: -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-99707


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.