Learn More
Cayman Chemical Phenylmethylsulfonyl fluoride
SDP
Supplier:  Cayman Chemical 143335 G
Encompass_Preferred
A nonspecific, irreversible inhibitor of serine proteases and other enzymes, including acetylcholinesterase, palmityl coenzyme A deacylase, arylsulfatase A, chymotrypsin, and trypsin; also known to block anandamide metabolism, which produces CB effects in mice, including antinociception, hypothermia, and immobility (ED50s = 86, 224, and 206 mg/kg, respectively)
Catalog No. 50-295-4023