Learn More
Crescent Chemical Co Inc PHENYLMETHYLSULFONYL FLUORIDE
SDP
Supplier:  Crescent Chemical Co Inc SE32395.02
Encompass_Preferred
Phenylmethylsulfonyl fluoride research grade Inhibits trypsin and chymotrypsin (1). Non-inhibitory to cholineesterase. Less toxic than diisopropylfluorophosphate (2). Inactivation of PMSF in buffer (3). Size - 5G Storage Conditions - +15 °C TO +30 °C Catalog Numb
Catalog No. 50-248-009