Learn More
Medchemexpress LLC Phenylphosphinic aci 100g | 1779-48-2 | 142.09 g/mol | C6H7O2P | 100 G
SDP
Supplier:  Medchemexpress LLC HY116210100G
Encompass_Preferred
Phenylphosphinic acid (CAS 1779-48-2) is an organophosphorus reagent used for surface modification and materials synthesis. It binds to metal oxide surfaces to functionalize interfaces and is used in nanoparticle preparation and sol-gel synthesis. Molecular formula C6H7O2P; molecular weight 142.09 g/mol.
  • Binds to metal oxide surfaces for modification and functionalization.
  • Suitable as a building block in nanoparticle and sol-gel synthesis.
  • Provides phosphorus-containing anchoring groups for inorganic substrates.
  • Offered in multiple pack sizes to support lab-scale and preparative work.
Catalog No. 50-002-29796