Learn More
Medchemexpress LLC Phenylphosphinic aci 500g | 1779-48-2 | 142.09 g/mol | C6H7O2P | 500 G
SDP
Supplier:  Medchemexpress LLC HY116210500G
Encompass_Preferred
Phenylphosphinic acid is an aromatic phosphinic acid used to modify and functionalize metal oxide surfaces and as a reagent in nanoparticle and sol-gel synthesis. It is typically supplied as a white powder of high purity for laboratory use.
  • Useful for surface modification of metal oxides.
  • Applicable to nanoparticle and sol-gel syntheses.
  • White powder, easy to handle and weigh.
  • High purity suitable for research applications.
  • Available in common lab-scale pack sizes for convenient use.
Catalog No. 50-002-27320