Learn More
Cayman Chemical Phenylpiracetam 5mg
SDP
Supplier:  Cayman Chemical 138975 MG
Encompass_Preferred
A nootropic derivative of the GABAergic agent piracetam that has been evaluated for its efficacy in reducing neurological deficits related to cerebral ischemia, epilepsy, and neurobehavioral disorders; intended for research and forensic purposes only
Catalog No. 50-295-4035