Learn More
AdipoGen Phenylpropargyl ether
SDP
Supplier:  AdipoGen CDXP0124G001
Encompass_Preferred
Chemical. CAS 13610-02-1. Formula C9H8O. MW 132.2. Synthetic. Aromatic terminal alkynes. Building block for synthesis. The terminal alkyne functionality has a wide range of applications.
Catalog No. 50-256-0243