Learn More
Medchemexpress LLC Phenylpropiolic acid | 637-44-5 | 146.15 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00731910G
Encompass_Preferred
Phenylpropiolic acid is an endogenous metabolite and a click chemistry reagent. It contains an Alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Endogenous metabolite
  • Click chemistry reagent
  • Contains Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition with Azide groups
Catalog No. 50-004-60349