Learn More
Medchemexpress LLC Phenyltrimethylammonium chloride | 138-24-9 | MFCD00011790 | ≥98.0% | 171.67 g/mol | C9H14ClN | 100 G
SDP
Supplier:  Medchemexpress LLC HYW016923100G
Encompass_Preferred
N,N,N-Trimethylbenzenaminium chloride is a quaternary ammonium salt (phenyltrimethylammonium chloride, CAS 138-24-9) supplied as a solid reagent for laboratory research and organic synthesis. It is used in life-science and synthetic chemistry workflows.
  • High purity (≥98.0%).
  • Chemical formula C9H14ClN; molecular weight 171.67 g/mol.
  • Solid form with a pale purple to purple appearance.
  • Suitable as a reagent in organic synthesis and research applications.
  • Available in laboratory quantities (25 g-500 g) for routine use.
  • For research use only; not for human or clinical use.
Catalog No. 50-003-02023