Learn More
U.S. Pharmacopeia Phenytoin, 57-41-0, MFCD00005264, 200 mg
Supplier:  U.S. Pharmacopeia 1535008
Encompass
Empirical Formula (Hill Notation): C15H12N2O2, Molecular Weight: 252.27, mp: 293-295 °C, Synonym: 5,5-Diphenyl-2,4-imidazolidinedione, 5,5-Diphenylhydantoin, Phenytoin.
Catalog No. NC1126466