Learn More
Medchemexpress LLC Phenytoin (5,5-Diphenylhydantoin) | 57-41-0 | 99.95% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB044810MM/1ML
Encompass_Preferred
Phenytoin (5,5-Diphenylhydantoin) is a potent blocker of Voltage-gated Na+ channels (VGSCs). It exhibits antiepileptic activity and has been shown to reduce breast tumor growth and metastasis in mice.
  • Potent voltage-gated Na+ channels (VGSCs) blocker.
  • Possesses antiepileptic activity, effective against partial seizures and generalized tonic-clonic seizures.
  • Protects against seizures by voltage-dependent block of voltage-gated sodium channels.
  • Inhibitory potency is strongly use-dependent.
  • Slow onset of sodium channel blocking.
  • Selectively inhibits pathological hyperexcitability in epilepsy.
  • Also blocks persistent sodium current.
  • Class 1b antiarrhythmic.
  • Reduces MDA-MB-231 cells in mice.
Catalog No. 50-004-06787