Learn More
Cayman Chemical PhenytoIn-d10 10mg
SDP
Supplier:  Cayman Chemical 3324510 MG
Encompass_Preferred
An internal standard for the quantification of phenytoin by GC- or LC-MS
Catalog No. 50-294-7856