Learn More
Medchemexpress LLC Phenytoin sodium | 630-93-3 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0448A10MM/1ML
Encompass_Preferred
Phenytoin sodium is a potent blocker of Voltage-gated Na+ channels (VGSCs). It demonstrates antiepileptic activity and has been observed to reduce breast tumor growth and metastasis in mice.
  • Potent voltage-gated Na+ channels blocker
  • Exhibits antiepileptic activity for partial and generalized tonic-clonic seizures
  • Protects against seizures by voltage-dependent block of voltage-gated sodium channels
  • Shows low affinity for resting sodium channels
  • Greater binding and block occur in depolarized neurons with open/inactivated channels
  • Displays strong use-dependent inhibitory potency
  • Selectively inhibits pathological hyperexcitability
  • Blocks persistent sodium current
  • Acts as a Class 1b antiarrhythmic
  • Reduces breast tumor growth and metastasis in mice
  • Can induce a model of gingival hyperplasia in mice
Catalog No. 50-004-07828