Learn More
Medchemexpress LLC HY-108637 10mg Medchemexpress, PhiKan 083 CAS:880813-36-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10863710MG
Encompass_Preferred
Medchemexpress, HY-108637 10mg PhiKan 083 CAS:880813-36-5 PhiKan 083 is a carbazole derivative, which binds to the surface cavity and stabilizes Y220C (a p53 mutant), with a Kd of 167 μM, and a relative binding affinity (Kd) of 150 μM in Ln229 cells[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8599