Learn More
Medchemexpress LLC Philanthotoxin 74 dihydrochloride | 1227301-51-0 | 99.7% | 507.54 g·mol⁻¹ | C24H44Cl2N4O3 | 10 MG

Supplier: Medchemexpress LLC HY104020A10MG
Philanthotoxin 74 dihydrochloride is a research-use AMPA receptor antagonist that inhibits GluR3 and GluR1 with reported IC50s of 263 nM and 296 nM. It is supplied in solid and solution formats for in vitro pharmacology and electrophysiology studies, with documented purity, solubility, and handling information.
- Research-use reagent; not for human or clinical use.
- Subtype-selective AMPAR antagonist; inhibits GluR3 (IC50 = 263 nM) and GluR1 (IC50 = 296 nM).
- Purity 99.72%.
- Molecular weight 507.54 g·mol⁻¹.
- CAS number 1227301-51-0.
- Soluble in DMSO (100 mg/mL) and water (50 mg/mL).
- Storage at 4°C, sealed, away from moisture.
- Available in solid and solution pack sizes including 1 mg, 5 mg, and 10 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.