Learn More
Selleck Chemical LLC Phlorizin 500mg 60-81-1 Phloridzin
SDP
Supplier:  Selleck Chemical LLC S2343500MG
Encompass_Preferred
Phlorizin (Phloridzin) is a dihydrochalcone found in the bark of pear (Pyrus communis), apple, cherry and other fruit trees. Phlorizin is a non-selective SGLT inhibitor with Ki of 300 nM and 39 nM for hSGLT1 and hSGLT2, respectively. Phlorizin is also a Na+/K+-ATPase inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2304