Learn More
Phloroglucinol Reagent, 0.7% w/v, Reagents
Phloroglucinol Reagent, 0.7% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$24.50
Chemical Identifiers
| CAS | 7732-18-5 , 67-56-1 , 7647-01-0 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | 1,3,5-Trihydroxybenzene Dihydrate, 1,3,5-Benzenetriol Dihydrate |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-334-407
|
Reagents Holdings Llc
CP106700120C |
120 mL |
Each of 1 for $24.50
|
|
|||||
Chemical Identifiers
| 7732-18-5 , 67-56-1 , 7647-01-0 | |
| 18.02 | |
| 1,3,5-Trihydroxybenzene Dihydrate, 1,3,5-Benzenetriol Dihydrate | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| Yellow to Orange | |
| 80°C | |
| 1,3,5-Trihydroxybenzene Dihydrate, 1,3,5-Benzenetriol Dihydrate | |
| O | |
| 18.02 | |
| Reagent | |
| 0.9954 | |
| Non-sterile | |
| Phloroglucinol Reagent, 0.7% w/v |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.