Learn More
Electron Microscopy Sciences Phloxine B, Certified, C.I. DcPh-6 25 GR

Supplier: Electron Microscopy Sciences 19350
(Acid Red 92; Cyanosine; Eosin 10B; Phloxine TA, N, BP Super, RB, TB, or BB)
CAS #18472-87-2 C₂0H₂O5Cl₄Br₄Na₂ F.W. 829.64
Solubility: 10.5% Water; 5.0% Alcohol; 9.0% Cellosolve; 4.5% Glycol; 0.0% Xylene
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.