Learn More
ATCC Phocaeicola vulgatus
Supplier:  ATCC 8482
Encompass
Phocaeicola vulgatus is a whole-genome sequenced bacterial type strain that has applications in media and sterility testing. This strain is recommended as a test microorganism for USP <71> Sterility Tests.
Catalog No. 50-238-4295