Learn More
Electron Microscopy Sciences Pholoxine B Stock 1% Aqueous 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2676302
Encompass_Preferred
Pholoxine B Stock in a 1% aqueous solution for Eosin-Phloxine Counterstain.
Catalog No. 50-319-10