Learn More
Cayman Chemical PhomopsInamIn 100ug
SDP
Supplier:  Cayman Chemical 33588100 UG
Encompass_Preferred
An inhibitor of microtubule polymerization; inhibits rate and extent of polymerization of tubulin isolated from ovine brain (IC50s = 0.53 and 0.59 µM, respectively)
Catalog No. 50-294-7829