Learn More
Supply Solutions PhosSTOP™ 10 tablets
Supplier:  Supply Solutions SIGMA 4906845001
Encompass
PhosSTOP tablets are a proprietary blend of phosphatase inhibitors, formulated as a ready-to-use tablet which dissolves quickly in aqueous solutions (buffers), including buffers containing formalin (for formalin fixation).
Catalog No. NC0922733