Learn More
Enzo Life Sciences Phorbol 12,13-dibutyrate (5mg). CAS: 37558-16-0
SDP
Supplier:  Enzo Life Sciences BMLPE1350005
Encompass_Preferred
Widely used PKC activator (10-100ng/ml). Less potent than phorbol 12-.myristate 13-acettate (PMA; TPA) (BML-PE160), but also less hydrophobic. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (20mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1399