Learn More
Medchemexpress LLC HY-18739 1mg Medchemexpress, Phorbol 12-myristate 13-acetate CAS:16561-29-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY187391MG
Encompass_Preferred
Medchemexpress, HY-18739 1mg Phorbol 12-myristate 13-acetate CAS:16561-29-8 Phorbol 12-myristate 13-acetate (PMA), a phorbol ester, is a dual SphK and protein kinase C (PKC) activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9269