Learn More
Selleck Chemical LLC Phorbol 1213-dibutyrate E7637-1MG
SDP
Supplier:  Selleck Chemical LLC E76371MG
Encompass_Preferred
Phorbol 12 13-dibutyrate (Phorbol dibutyrate) is a PKC activator and a potent skin tumor promoter
Catalog No. 50-000-43161