Learn More
AG Scientific Inc Phosphatase Inhib II
Supplier:  AG Scientific Inc P1518SOLIDVIAL
Encompass_Preferred
"Phosphatase Inhibitor Cocktail II, Solid is a mixture of five phosphatase inhibitors which has been optimized for the inhibition of protein tyrosine phosphatases, acid and alkaline phosphatases. Phosphatase Inhibitor Cocktail protects phosphoproteins from dephosphorylation during lysis and extraction procedures from mammalian cells or tissues and for immunoprecipitation and kinase assays. Phosphatase inhibitor cocktails cover the widest range of phosphatases to ensure the protection of proteins from dephosphorylation. Components: ​​Imidazole (200 mM) Sodium fluoride (100 mM) Sodium Molybdate (115 mM) Sodium Orthovanadate (100 mM) Sodium tartrate, dihyrdrate (400 mM)"
Catalog No. 50-252-2608