Learn More
Sigma Aldrich Fine Chemicals Biosciences Phosphatase Inhibitor Cocktail 2 aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P57261ML
Encompass
Phosphatase Inhibitor Cocktail 2 aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Catalog No. 50-165-7375