Learn More
Hach Company Phosphate Standard Solution, 50 mg/L as PO₄ (NIST), 16/pk - 10-mL Voluette« Ampules
Supplier:  Hach Company 17110
Encompass
For accuracy checks in phosphate (phosphorus) determinations
Catalog No. 50-199-4187