Learn More
Research Products International Corp Phosphatase Inhibitor Cocktail III, 1 Vial

Supplier: Research Products International Corp P521041
Lyophilized blend of four phosphatase inhibitors for use with animal tissue estracts. Reconstitute with 1ml water to produce a 1 ml concentrate.
Contains:
Sodium Orthovanadate
Sodium Pyrophosphate, decahydrate
B-Glycerophosphate
Specificity:
Protein Tyrosine
Phosphatases
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.