Learn More
Medchemexpress LLC Phosphine, [1,1'-biphenyl]-2-ylbis(1,1-dimethylethyl)- | 224311-51-7 | 298.41 | 10 G

Supplier: Medchemexpress LLC HYW00732310G
JohnPhos is a biochemical reagent intended for life science-related research. It can be utilized as a biological material or an organic compound. This product is for research use only and not sold to patients.
- Suitable for various life science research purposes
- Comes with a purity data sheet and other documentation
- Appearance: solid, white to off-white
- Soluble in DMSO at 10 mg/mL (33.51 mM)
- Recommended storage at 4°C under nitrogen
- In solvent, storage is recommended at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.