Learn More
ABclonal Technology Phospho-ACC1/ACC2-S79 Rabbit
SDP
Supplier:  ABclonal Technology AP1410PM100UL
Encompass_Preferred
Acetyl-CoA carboxylase (ACC) is a complex multifunctional enzyme system ACC is a biotin-containing enzyme which catalyzes the carboxylation of acetyl-CoA to malonyl-CoA the rate-limiting step in fatty acid synthesis There are two ACC forms alpha and beta encoded by two different genes ACC-alpha is highly enriched in lipogenic tissues The enzyme is under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA Multiple alternatively spliced transcript variants divergent in the 5 sequence and encoding distinct isoforms have been found for this gene
Catalog No. 50-000-14043