Learn More
Meso Scale Discovery PHOSPHO-AKT SER473 KIT, 5 plates

Supplier: Meso Scale Discovery K150MND2
The MESO SCALE DISCOVERY K150MND-2 PHOSPHO-AKT SER473 KIT enables quantitative detection of phosphorylated Akt1 at serine 473 using an electrochemiluminescence-based immunoassay. This kit is essential for research on cell growth, survival, cancer, and other conditions, offering a rapid and efficient approach to measure protein levels from small sample volumes. It accommodates various sample types, including human and animal cell lysates, making it versatile for different biological studies.
- Kit size: 5 plates
- Target analyte: Phospho-Akt (Ser473)
- Phosphorylation site: Serine 473
- Detection method: Electrochemiluminescence (MSD SULFO-TAG™ labels)
- Assay principle: Sandwich immunoassay
- Sample compatibility: Human, mouse, rat, non-human primate cell lysates and tissue lysates
- Regulatory status: For research use only
- Diagnostic use: Not for use in diagnostic procedures
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.