Learn More
Medchemexpress LLC Phospho-AKT1 (Ser473) antibody (YA228) | 100 UL

Supplier: Medchemexpress LLC HYP80788100UL
Phospho-AKT1 (Ser473) Antibody (YA228) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed to target Phospho-AKT1 (Ser473). This antibody is suitable for western blot applications and reacts with human, mouse, and rat samples.
- Host: Rabbit
- Reactivity: Human, mouse, rat
- Application: Western blot
- Clonality: Monoclonal
- Isotype: IgG
- Conjugation: Non-conjugated
- Immunogen: Phospho-AKT1 (Ser473)
- Molecular weight: 56 kDa
- Formulation: Supplied in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol and 0.05% BSA, with 0.01% Sodium azide preservative.
- Storage: Stored at -20°C for 1 year; avoid repeated freeze / thaw cycles.
- Shipping: Shipped with blue ice.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.